Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001037872.2 | NP_001032961.1 | 14 - 15 | 2587 | 65 |
NM_001321454.1 | NP_001308383.1 | 15 - 16 | 2698 | 65 | |
NM_001321455.1 | NP_001308384.1 | 15 - 16 | 2744 | 65 | |
NM_001321458.1 | NP_001308387.1 | 15 - 16 | 2714 | 65 | |
NM_001321459.1 | NP_001308388.1 | 15 - 16 | 2790 | 65 | |
NM_001321460.1 | NP_001308389.1 | 15 - 16 | 2787 | 65 | |
NM_016316.3 | NP_057400.1 | 14 - 15 | 2590 | 65 | |
NR_135649.1 | 15 - 16 | 2787 | 65 | ||
NR_135650.1 | 15 - 16 | 2744 | 65 | ||
NR_135651.1 | 14 - 15 | 2587 | 65 | ||
NR_135652.1 | 14 - 15 | 2590 | 65 | ||
NR_135653.1 | 14 - 15 | 2590 | 65 | ||
XM_011511341.2 | XP_011509643.1 | 8 - 9 | 1207 | 65 | |
XM_017004309.1 | XP_016859798.1 | 15 - 16 | 3111 | 65 | |
XM_017004310.1 | XP_016859799.1 | 15 - 16 | 2477 | 65 | |
XM_017004311.1 | XP_016859800.1 | 15 - 16 | 2477 | 65 | |
XM_017004312.1 | XP_016859801.1 | 15 - 16 | 2690 | 65 | |
XM_017004313.1 | XP_016859802.1 | 14 - 15 | 4316 | 65 | |
XM_017004314.1 | XP_016859803.1 | 11 - 12 | 1967 | 65 | |
XM_017004315.1 | XP_016859804.1 | 11 - 12 | 1951 | 65 | |
GenBank mRNA | AB047646.1 | - | 14 - 15 | 2512 | 65 |
AF151538.1 | - | 14 - 15 | 2543 | 65 | |
AF206019.1 | - | 14 - 15 | 2552 | 65 | |
AF357886.1 | - | 14 - 15 | 2508 | 65 | |
AJ131720.1 | - | 6 - 7 | 909 | 65 | |
AK025176.1 | - | 6 - 7 | 869 | 65 | |
AK307719.1 | - | 14 - 15 | 2501 | 65 | |
BC037734.1 | - | 15 - 16 | 2725 | 65 | |
BC130411.1 | - | 13 - 14 | 2358 | 65 |