Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001037872.2 | NP_001032961.1 | 3 - 4 | 437 | 119 |
NM_001321454.1 | NP_001308383.1 | 3 - 4 | 437 | 119 | |
NM_001321458.1 | NP_001308387.1 | 3 - 4 | 437 | 119 | |
NM_001321459.1 | NP_001308388.1 | 3 - 4 | 437 | 119 | |
NM_001321460.1 | NP_001308389.1 | 3 - 4 | 437 | 119 | |
NM_016316.3 | NP_057400.1 | 3 - 4 | 437 | 119 | |
NR_135649.1 | 3 - 4 | 437 | 119 | ||
NR_135651.1 | 3 - 4 | 437 | 119 | ||
NR_135652.1 | 3 - 4 | 437 | 119 | ||
NR_135653.1 | 3 - 4 | 437 | 119 | ||
XM_017004309.1 | XP_016859798.1 | 3 - 4 | 850 | 119 | |
XM_017004310.1 | XP_016859799.1 | 3 - 4 | 216 | 119 | |
XM_017004311.1 | XP_016859800.1 | 3 - 4 | 216 | 119 | |
XM_017004312.1 | XP_016859801.1 | 3 - 4 | 432 | 119 | |
XM_017004313.1 | XP_016859802.1 | 3 - 4 | 2166 | 119 | |
GenBank mRNA | AB047646.1 | - | 3 - 4 | 362 | 119 |
AF151538.1 | - | 3 - 4 | 390 | 119 | |
AF206019.1 | - | 3 - 4 | 399 | 119 | |
AF357886.1 | - | 3 - 4 | 355 | 119 | |
AK002087.1 | - | 1 - 2 | 299 | 119 | |
AK307719.1 | - | 3 - 4 | 348 | 119 | |
BC130411.1 | - | 2 - 3 | 208 | 119 | |
DR000409.1 | - | 3 - 4 | 328 | 119 | |
HY010913.1 | - | 3 - 4 | 438 | 119 |