Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005813.4 | NP_005804.1 | 12 - 13 | 2332 | 64 |
XM_005264237.3 | XP_005264294.1 | 13 - 14 | 2605 | 64 | |
XM_005264238.3 | XP_005264295.1 | 12 - 13 | 1581 | 64 | |
XM_017003741.1 | XP_016859230.1 | 14 - 15 | 2610 | 64 | |
XM_017003742.1 | XP_016859231.1 | 13 - 14 | 1649 | 64 | |
XM_017003743.1 | XP_016859232.1 | 10 - 11 | 1331 | 64 | |
XM_017003744.1 | XP_016859233.1 | 9 - 10 | 1079 | 64 | |
GenBank mRNA | AB015982.2 | - | 12 - 13 | 2332 | 64 |
AK027585.1 | - | 9 - 10 | 1141 | 64 | |
AK122883.1 | - | 5 - 6 | 2006 | 64 |