Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005813.4 | NP_005804.1 | 5 - 6 | 1460 | 61 |
XM_005264237.3 | XP_005264294.1 | 6 - 7 | 1733 | 61 | |
XM_005264238.3 | XP_005264295.1 | 5 - 6 | 709 | 61 | |
XM_017003741.1 | XP_016859230.1 | 7 - 8 | 1738 | 61 | |
XM_017003742.1 | XP_016859231.1 | 6 - 7 | 777 | 61 | |
XM_017003743.1 | XP_016859232.1 | 3 - 4 | 459 | 61 | |
XM_017003744.1 | XP_016859233.1 | 2 - 3 | 207 | 61 | |
GenBank mRNA | AB015982.2 | - | 5 - 6 | 1460 | 61 |
AK027585.1 | - | 2 - 3 | 269 | 61 | |
BC012820.2 | - | 5 - 6 | 1419 | 61 | |
BC030706.2 | - | 5 - 6 | 1405 | 61 |