Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001270660.1 | NP_001257589.1 | 12 - 13 | 1236 | 87 |
NM_001270661.1 | NP_001257590.1 | 10 - 11 | 1065 | 87 | |
NM_005885.3 | NP_005876.2 | 13 - 14 | 1380 | 87 | |
XM_011513932.2 | XP_011512234.1 | 13 - 14 | 1138 | 87 | |
XM_011513934.1 | XP_011512236.1 | 13 - 14 | 1341 | 87 | |
XM_011513936.2 | XP_011512238.1 | 13 - 14 | 1341 | 87 | |
XM_011513937.2 | XP_011512239.1 | 3 - 4 | 722 | 87 | |
XM_017008944.1 | XP_016864433.1 | 13 - 14 | 1341 | 87 | |
GenBank mRNA | AB011169.1 | - | 13 - 14 | 1303 | 87 |
AK296585.1 | - | 10 - 11 | 988 | 87 | |
AK300034.1 | - | 12 - 13 | 1172 | 87 | |
AK314686.1 | - | 2 - 3 | 233 | 87 | |
BC046148.1 | - | 5 - 6 | 297 | 87 | |
BC136461.1 | - | 13 - 14 | 1252 | 87 | |
BC136462.1 | - | 13 - 14 | 1252 | 87 | |
BC142679.1 | - | 13 - 14 | 1231 | 87 | |
BC142694.1 | - | 13 - 14 | 1231 | 87 |