Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001270660.1 | NP_001257589.1 | 19 - 20 | 2082 | 63 |
NM_001270661.1 | NP_001257590.1 | 17 - 18 | 1911 | 63 | |
NM_005885.3 | NP_005876.2 | 20 - 21 | 2226 | 63 | |
XM_011513932.2 | XP_011512234.1 | 20 - 21 | 1984 | 63 | |
XM_011513934.1 | XP_011512236.1 | 20 - 21 | 2187 | 63 | |
XM_011513937.2 | XP_011512239.1 | 10 - 11 | 1568 | 63 | |
GenBank mRNA | AB011169.1 | - | 20 - 21 | 2149 | 63 |
AF009301.1 | - | 7 - 8 | 845 | 63 | |
AK296585.1 | - | 17 - 18 | 1834 | 63 | |
AK300034.1 | - | 19 - 20 | 2018 | 63 | |
AK314686.1 | - | 9 - 10 | 1079 | 63 | |
BC046148.1 | - | 12 - 13 | 1143 | 63 | |
BC136461.1 | - | 20 - 21 | 2098 | 63 | |
BC136462.1 | - | 20 - 21 | 2098 | 63 | |
BC142679.1 | - | 20 - 21 | 2077 | 63 | |
BC142694.1 | - | 20 - 21 | 2077 | 63 |