Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001270660.1 | NP_001257589.1 | 20 - 21 | 2267 | 75 |
NM_001270661.1 | NP_001257590.1 | 18 - 19 | 2096 | 75 | |
NM_005885.3 | NP_005876.2 | 21 - 22 | 2411 | 75 | |
XM_011513932.2 | XP_011512234.1 | 21 - 22 | 2169 | 75 | |
XM_011513934.1 | XP_011512236.1 | 21 - 22 | 2372 | 75 | |
XM_011513937.2 | XP_011512239.1 | 11 - 12 | 1753 | 75 | |
GenBank mRNA | AB011169.1 | - | 21 - 22 | 2334 | 75 |
AF009301.1 | - | 8 - 9 | 1030 | 75 | |
AK296585.1 | - | 18 - 19 | 2019 | 75 | |
AK300034.1 | - | 20 - 21 | 2203 | 75 | |
AK314686.1 | - | 10 - 11 | 1264 | 75 | |
BC046148.1 | - | 13 - 14 | 1328 | 75 | |
BC136461.1 | - | 21 - 22 | 2283 | 75 | |
BC136462.1 | - | 21 - 22 | 2283 | 75 | |
BC142679.1 | - | 21 - 22 | 2262 | 75 | |
BC142694.1 | - | 21 - 22 | 2262 | 75 |