Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001270660.1 | NP_001257589.1 | 6 - 7 | 883 | 68 |
NM_001270661.1 | NP_001257590.1 | 4 - 5 | 712 | 68 | |
NM_005885.3 | NP_005876.2 | 7 - 8 | 1027 | 68 | |
XM_011513932.2 | XP_011512234.1 | 7 - 8 | 785 | 68 | |
XM_011513934.1 | XP_011512236.1 | 7 - 8 | 988 | 68 | |
XM_011513936.2 | XP_011512238.1 | 7 - 8 | 988 | 68 | |
XM_017008944.1 | XP_016864433.1 | 7 - 8 | 988 | 68 | |
GenBank mRNA | AB011169.1 | - | 7 - 8 | 950 | 68 |
AK000970.1 | - | 7 - 8 | 942 | 68 | |
AK296585.1 | - | 4 - 5 | 635 | 68 | |
AK300034.1 | - | 6 - 7 | 819 | 68 | |
BC136461.1 | - | 7 - 8 | 899 | 68 | |
BC136462.1 | - | 7 - 8 | 899 | 68 | |
BC142679.1 | - | 7 - 8 | 878 | 68 | |
BC142694.1 | - | 7 - 8 | 878 | 68 |