Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001270660.1 | NP_001257589.1 | 8 - 9 | 980 | 109 |
NM_001270661.1 | NP_001257590.1 | 6 - 7 | 809 | 109 | |
NM_005885.3 | NP_005876.2 | 9 - 10 | 1124 | 109 | |
XM_011513932.2 | XP_011512234.1 | 9 - 10 | 882 | 109 | |
XM_011513934.1 | XP_011512236.1 | 9 - 10 | 1085 | 109 | |
XM_011513936.2 | XP_011512238.1 | 9 - 10 | 1085 | 109 | |
XM_017008944.1 | XP_016864433.1 | 9 - 10 | 1085 | 109 | |
GenBank mRNA | AB011169.1 | - | 9 - 10 | 1047 | 109 |
AK000970.1 | - | 9 - 10 | 1039 | 109 | |
AK296585.1 | - | 6 - 7 | 732 | 109 | |
AK300034.1 | - | 8 - 9 | 916 | 109 | |
BC046148.1 | - | 1 - 2 | 41 | 109 | |
BC136461.1 | - | 9 - 10 | 996 | 109 | |
BC136462.1 | - | 9 - 10 | 996 | 109 | |
BC142679.1 | - | 9 - 10 | 975 | 109 | |
BC142694.1 | - | 9 - 10 | 975 | 109 |