Assay Design



Gene Transcripts

Gene Symbol
DNAH6
Entrez Gene ID
1768
Gene Name
dynein axonemal heavy chain 6
Gene Aliases
DNHL1, Dnahc6, HL-2, HL2
Chromosome Location
Chr.2: 84459588 - 84819589 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001370.1 NP_001361.1 29 - 30 4655 80
XM_006711956.2 XP_006712019.1 29 - 30 4760 80
XM_011532649.2 XP_011530951.1 29 - 30 4609 80
XM_011532650.2 XP_011530952.1 29 - 30 4610 80
XM_011532652.1 XP_011530954.1 28 - 29 4529 80
XM_011532653.1 XP_011530955.1 29 - 30 4655 80
XM_011532654.1 XP_011530956.1 28 - 29 4388 80
XM_011532655.2 XP_011530957.1 25 - 26 3677 80
XM_011532657.1 XP_011530959.1 23 - 24 3416 80
XM_011532660.1 XP_011530962.1 29 - 30 4655 80
XM_017003521.1 XP_016859010.1 28 - 29 4529 80
XM_017003522.1 XP_016859011.1 24 - 25 4481 80
XM_017003523.1 XP_016859012.1 24 - 25 3648 80
XM_017003524.1 XP_016859013.1 29 - 30 4655 80
GenBank mRNA U61736.1 - 1 - 2 177 80

View Full Product Details