Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006391.2 | NP_006382.1 | 23 - 24 | 3045 | 124 |
GenBank mRNA | AF098799.1 | - | 23 - 24 | 3010 | 124 |
AK027487.1 | - | 3 - 4 | 549 | 124 | |
AK074908.1 | - | 16 - 17 | 2076 | 124 | |
AK312885.1 | - | 23 - 24 | 3016 | 124 | |
AL137335.1 | - | 2 - 3 | 415 | 124 | |
BC114929.1 | - | 23 - 24 | 2971 | 124 |