Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006391.2 | NP_006382.1 | 9 - 10 | 1180 | 91 |
GenBank mRNA | AF098799.1 | - | 9 - 10 | 1145 | 91 |
AK074908.1 | - | 2 - 3 | 211 | 91 | |
AK312885.1 | - | 9 - 10 | 1151 | 91 | |
BC114929.1 | - | 9 - 10 | 1106 | 91 |