Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001025598.1 | NP_001020769.1 | 9 - 10 | 1368 | 107 |
NM_001287600.1 | NP_001274529.1 | 8 - 9 | 1223 | 107 | |
NM_001287602.1 | NP_001274531.1 | 5 - 6 | 837 | 107 | |
NM_181720.2 | NP_859071.2 | 9 - 10 | 1368 | 107 | |
XM_005245070.2 | XP_005245127.1 | 9 - 10 | 1368 | 107 | |
XM_005245073.3 | XP_005245130.1 | 7 - 8 | 1121 | 107 | |
XM_011509391.2 | XP_011507693.1 | 8 - 9 | 1199 | 107 | |
GenBank mRNA | AK126163.1 | - | 5 - 6 | 837 | 107 |
AK160366.1 | - | 9 - 10 | 1343 | 107 | |
AL832852.1 | - | 7 - 8 | 803 | 107 | |
BC043387.1 | - | 8 - 9 | 1056 | 107 | |
BC053688.1 | - | 9 - 10 | 1064 | 107 |