Assay Design



Gene Transcripts

Gene Symbol
ARPP21
Entrez Gene ID
10777
Gene Name
cAMP regulated phosphoprotein 21
Gene Aliases
ARPP-21, R3HDM3, RCS, TARPP
Chromosome Location
Chr.3: 35638981 - 35798933 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025068.1 NP_001020239.1 4 - 5 447 68
NM_001025069.1 NP_001020240.1 4 - 5 456 68
NM_001267616.1 NP_001254545.1 4 - 5 1603 68
NM_001267617.1 NP_001254546.1 4 - 5 413 68
NM_001267618.1 NP_001254547.1 3 - 4 544 68
NM_001267619.1 NP_001254548.1 3 - 4 378 68
NM_016300.4 NP_057384.2 4 - 5 430 68
NM_198399.1 NP_938409.1 3 - 4 438 68
XM_005264811.2 XP_005264868.1 4 - 5 459 68
XM_006712943.2 XP_006713006.1 4 - 5 459 68
XM_006712944.2 XP_006713007.1 4 - 5 459 68
XM_011533298.2 XP_011531600.1 4 - 5 459 68
XM_011533299.2 XP_011531601.1 4 - 5 1003 68
XM_011533300.2 XP_011531602.1 4 - 5 622 68
XM_011533301.2 XP_011531603.1 4 - 5 453 68
XM_011533302.2 XP_011531604.1 5 - 6 545 68
XM_011533303.2 XP_011531605.1 5 - 6 1095 68
XM_017005574.1 XP_016861063.1 4 - 5 549 68
XM_017005575.1 XP_016861064.1 5 - 6 12182 68
XM_017005576.1 XP_016861065.1 4 - 5 1003 68
XM_017005577.1 XP_016861066.1 4 - 5 453 68
XM_017005578.1 XP_016861067.1 4 - 5 429 68
XM_017005579.1 XP_016861068.1 4 - 5 1003 68
XM_017005580.1 XP_016861069.1 5 - 6 1095 68
XM_017005581.1 XP_016861070.1 5 - 6 12182 68
XM_017005582.1 XP_016861071.1 4 - 5 1003 68
XM_017005583.1 XP_016861072.1 4 - 5 453 68
XM_017005584.1 XP_016861073.1 4 - 5 1003 68
XM_017005585.1 XP_016861074.1 4 - 5 1003 68
XM_017005586.1 XP_016861075.1 4 - 5 459 68
XM_017005587.1 XP_016861076.1 4 - 5 622 68
XM_017005588.1 XP_016861077.1 5 - 6 1095 68
XM_017005589.1 XP_016861078.1 5 - 6 545 68
XM_017005590.1 XP_016861079.1 5 - 6 12182 68
XM_017005591.1 XP_016861080.1 4 - 5 1003 68
XM_017005592.1 XP_016861081.1 4 - 5 1003 68
XM_017005593.1 XP_016861082.1 4 - 5 549 68
XM_017005594.1 XP_016861083.1 4 - 5 453 68
XM_017005595.1 XP_016861084.1 5 - 6 1095 68
XM_017005596.1 XP_016861085.1 4 - 5 1003 68
XM_017005597.1 XP_016861086.1 4 - 5 1003 68
XM_017005598.1 XP_016861087.1 5 - 6 1095 68
XM_017005599.1 XP_016861088.1 4 - 5 1003 68
XM_017005600.1 XP_016861089.1 4 - 5 622 68
XM_017005601.1 XP_016861090.1 4 - 5 453 68
XM_017005602.1 XP_016861091.1 4 - 5 1003 68
XM_017005603.1 XP_016861092.1 4 - 5 622 68
XM_017005604.1 XP_016861093.1 4 - 5 453 68
XM_017005605.1 XP_016861094.1 5 - 6 1095 68
XM_017005606.1 XP_016861095.1 5 - 6 545 68
XM_017005607.1 XP_016861096.1 4 - 5 1003 68
XM_017005608.1 XP_016861097.1 4 - 5 1003 68
XM_017005609.1 XP_016861098.1 5 - 6 1095 68
XM_017005610.1 XP_016861099.1 4 - 5 1003 68
XM_017005611.1 XP_016861100.1 4 - 5 1003 68
XM_017005612.1 XP_016861101.1 4 - 5 1003 68
GenBank mRNA AF112220.1 - 3 - 4 403 68
AK055064.1 - 4 - 5 456 68
AK289868.1 - 4 - 5 450 68
AK294484.1 - 3 - 4 391 68
AV718064.1 - 4 - 5 447 68
AV718166.1 - 4 - 5 456 68
BC017805.1 - 3 - 4 438 68
BC036399.1 - 4 - 5 418 68
BC041385.1 - 4 - 5 1191 68
BC051828.1 - 4 - 5 430 68
BC066651.1 - 2 - 3 957 68
BG535216.1 - 3 - 4 439 69

View Full Product Details