Assay Design



Gene Transcripts

Gene Symbol
ARPP21
Entrez Gene ID
10777
Gene Name
cAMP regulated phosphoprotein 21
Gene Aliases
ARPP-21, R3HDM3, RCS, TARPP
Chromosome Location
Chr.3: 35638981 - 35798933 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001267617.1 NP_001254546.1 16 - 17 2095 120
NM_001267619.1 NP_001254548.1 16 - 17 2120 120
NM_016300.4 NP_057384.2 17 - 18 2169 120
XM_005264811.2 XP_005264868.1 18 - 19 2303 120
XM_006712943.2 XP_006713006.1 17 - 18 2201 120
XM_006712944.2 XP_006713007.1 16 - 17 2141 120
XM_011533298.2 XP_011531600.1 18 - 19 2306 120
XM_011533299.2 XP_011531601.1 18 - 19 2850 120
XM_011533300.2 XP_011531602.1 18 - 19 2469 120
XM_011533301.2 XP_011531603.1 18 - 19 2300 120
XM_011533302.2 XP_011531604.1 19 - 20 2392 120
XM_011533303.2 XP_011531605.1 19 - 20 2942 120
XM_017005574.1 XP_016861063.1 18 - 19 2396 120
XM_017005575.1 XP_016861064.1 19 - 20 14029 120
XM_017005576.1 XP_016861065.1 18 - 19 2847 120
XM_017005577.1 XP_016861066.1 18 - 19 2297 120
XM_017005578.1 XP_016861067.1 18 - 19 2273 120
XM_017005579.1 XP_016861068.1 18 - 19 2847 120
XM_017005580.1 XP_016861069.1 19 - 20 2939 120
XM_017005581.1 XP_016861070.1 19 - 20 14026 120
XM_017005582.1 XP_016861071.1 18 - 19 2844 120
XM_017005583.1 XP_016861072.1 18 - 19 2294 120
XM_017005584.1 XP_016861073.1 17 - 18 2790 120
XM_017005585.1 XP_016861074.1 17 - 18 2787 120
XM_017005586.1 XP_016861075.1 17 - 18 2243 120
XM_017005587.1 XP_016861076.1 17 - 18 2406 120
XM_017005588.1 XP_016861077.1 18 - 19 2879 120
XM_017005589.1 XP_016861078.1 18 - 19 2329 120
XM_017005590.1 XP_016861079.1 18 - 19 13966 120
XM_017005591.1 XP_016861080.1 17 - 18 2748 120
XM_017005592.1 XP_016861081.1 17 - 18 2745 120
XM_017005593.1 XP_016861082.1 17 - 18 2291 120
XM_017005594.1 XP_016861083.1 17 - 18 2195 120
XM_017005595.1 XP_016861084.1 18 - 19 2837 120
XM_017005596.1 XP_016861085.1 17 - 18 2745 120
XM_017005597.1 XP_016861086.1 17 - 18 2742 120
XM_017005598.1 XP_016861087.1 18 - 19 2834 120
XM_017005599.1 XP_016861088.1 16 - 17 2688 120
XM_017005600.1 XP_016861089.1 16 - 17 2307 120
XM_017005601.1 XP_016861090.1 16 - 17 2138 120
XM_017005602.1 XP_016861091.1 16 - 17 2685 120
XM_017005603.1 XP_016861092.1 16 - 17 2304 120
XM_017005604.1 XP_016861093.1 16 - 17 2135 120
XM_017005605.1 XP_016861094.1 17 - 18 2777 120
XM_017005606.1 XP_016861095.1 17 - 18 2227 120
XM_017005607.1 XP_016861096.1 17 - 18 2705 120
XM_017005608.1 XP_016861097.1 19 - 20 2938 120
XM_017005609.1 XP_016861098.1 19 - 20 2970 120
XM_017005610.1 XP_016861099.1 16 - 17 2636 120
XM_017005611.1 XP_016861100.1 15 - 16 2576 120
XM_017005612.1 XP_016861101.1 16 - 17 2633 120
GenBank mRNA AK294484.1 - 16 - 17 2133 120
AL133109.1 - 10 - 11 1424 120
BC031106.1 - 5 - 6 981 120
BC036399.1 - 16 - 17 2100 120
BC051828.1 - 17 - 18 2169 120

View Full Product Details