Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001322253.1 | NP_001309182.1 | 17 - 18 | 2849 | 63 |
NM_001322256.1 | NP_001309185.1 | 15 - 16 | 2739 | 63 | |
NM_001322257.1 | NP_001309186.1 | 15 - 16 | 2641 | 63 | |
NM_015106.3 | NP_055921.2 | 16 - 17 | 2801 | 63 | |
XM_006713062.3 | XP_006713125.1 | 15 - 16 | 2548 | 63 | |
XM_017005987.1 | XP_016861476.1 | 15 - 16 | 2577 | 63 | |
XM_017005988.1 | XP_016861477.1 | 16 - 17 | 4137 | 63 | |
GenBank mRNA | AB018352.2 | - | 13 - 14 | 2206 | 63 |
AK023824.1 | - | 10 - 11 | 3001 | 63 | |
AK025049.1 | - | 5 - 6 | 647 | 63 | |
AK122683.1 | - | 15 - 16 | 2650 | 63 | |
AK304881.1 | - | 11 - 12 | 1937 | 63 | |
BC001474.2 | - | 9 - 10 | 1529 | 63 | |
BC024298.1 | - | 13 - 14 | 2165 | 63 |