Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001322253.1 | NP_001309182.1 | 13 - 14 | 2253 | 103 |
NM_001322256.1 | NP_001309185.1 | 11 - 12 | 2143 | 103 | |
NM_001322257.1 | NP_001309186.1 | 11 - 12 | 2051 | 103 | |
NM_015106.3 | NP_055921.2 | 12 - 13 | 2205 | 103 | |
XM_006713062.3 | XP_006713125.1 | 11 - 12 | 1952 | 103 | |
XM_017005987.1 | XP_016861476.1 | 11 - 12 | 1981 | 103 | |
XM_017005988.1 | XP_016861477.1 | 12 - 13 | 3541 | 103 | |
GenBank mRNA | AB018352.2 | - | 9 - 10 | 1610 | 103 |
AK122683.1 | - | 11 - 12 | 2060 | 103 | |
AK304881.1 | - | 7 - 8 | 1341 | 103 | |
AK308456.1 | - | 10 - 11 | 1742 | 103 | |
BC001474.2 | - | 5 - 6 | 933 | 103 | |
BC024298.1 | - | 9 - 10 | 1569 | 103 |