Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014289.3 | NP_055104.2 | 1 - 2 | 174 | 145 |
GenBank mRNA | AJ000388.1 | - | 1 - 2 | 165 | 145 |
AK312701.1 | - | 1 - 2 | 138 | 145 | |
DA863483.1 | - | 1 - 2 | 174 | 145 |