Assay Design



Gene Transcripts

Gene Symbol
CACNA2D2
Entrez Gene ID
9254
Gene Name
calcium voltage-gated channel auxiliary subunit alpha2delta 2
Gene Aliases
CACNA2D
Chromosome Location
Chr.3: 50362613 - 50504244 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005505.2 NP_001005505.1 10 - 11 1165 78
NM_001174051.2 NP_001167522.1 10 - 11 1165 78
NM_001291101.1 NP_001278030.1 10 - 11 961 78
NM_006030.3 NP_006021.2 10 - 11 1165 78
XM_005265581.4 XP_005265638.1 10 - 11 1039 78
XM_011534243.2 XP_011532545.1 10 - 11 1039 78
GenBank mRNA AB011130.1 - 10 - 11 992 78
AF040709.1 - 10 - 11 1171 78
AF042792.1 - 11 - 12 1152 78
AF042793.1 - 10 - 11 968 78
AJ251367.1 - 10 - 11 991 78
AJ251368.1 - 10 - 11 991 78
BC152438.1 - 10 - 11 992 78

View Full Product Details