Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001005505.2 | NP_001005505.1 | 9 - 10 | 1071 | 57 |
NM_001174051.2 | NP_001167522.1 | 9 - 10 | 1071 | 57 | |
NM_001291101.1 | NP_001278030.1 | 9 - 10 | 867 | 57 | |
NM_006030.3 | NP_006021.2 | 9 - 10 | 1071 | 57 | |
XM_005265581.4 | XP_005265638.1 | 9 - 10 | 945 | 57 | |
XM_011534243.2 | XP_011532545.1 | 9 - 10 | 945 | 57 | |
GenBank mRNA | AB011130.1 | - | 9 - 10 | 898 | 57 |
AF040709.1 | - | 9 - 10 | 1077 | 57 | |
AF042792.1 | - | 10 - 11 | 1058 | 57 | |
AF042793.1 | - | 9 - 10 | 874 | 57 | |
AJ251367.1 | - | 9 - 10 | 897 | 57 | |
AJ251368.1 | - | 9 - 10 | 897 | 57 | |
BC152438.1 | - | 9 - 10 | 898 | 57 |