Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018313.4 | NP_060783.3 | 26 - 27 | 4245 | 78 |
XM_017006753.1 | XP_016862242.1 | 29 - 30 | 5205 | 78 | |
XM_017006755.1 | XP_016862244.1 | 28 - 29 | 4709 | 78 | |
XM_017006756.1 | XP_016862245.1 | 28 - 29 | 4706 | 78 | |
XM_017006757.1 | XP_016862246.1 | 26 - 27 | 4432 | 78 | |
XM_017006758.1 | XP_016862247.1 | 26 - 27 | 4563 | 78 | |
XM_017006759.1 | XP_016862248.1 | 28 - 29 | 4634 | 78 | |
GenBank mRNA | AF177387.1 | - | 25 - 26 | 4290 | 78 |
AF197569.1 | - | 26 - 27 | 4227 | 78 | |
AF225870.1 | - | 24 - 25 | 4194 | 78 | |
AF225871.1 | - | 24 - 25 | 4075 | 78 | |
AK001507.1 | - | 2 - 3 | 317 | 78 | |
BC015323.1 | - | 3 - 4 | 581 | 78 | |
BC089409.1 | - | 4 - 5 | 662 | 78 | |
BC115009.1 | - | 25 - 26 | 4302 | 78 | |
BC115010.1 | - | 25 - 26 | 4231 | 78 | |
BC115011.1 | - | 25 - 26 | 78 |