Assay Design



Gene Transcripts

Gene Symbol
NCAN
Entrez Gene ID
1463
Gene Name
neurocan
Gene Aliases
CSPG3
Chromosome Location
Chr.19: 19211973 - 19252252 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004386.2 NP_004377.2 6 - 7 1175 62
GenBank mRNA AB210004.1 - 6 - 7 1119 62
AF026547.1 - 5 - 6 1077 62

View Full Product Details