Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017647.3 | NP_060117.3 | 17 - 18 | 2621 | 95 |
GenBank mRNA | AF327355.1 | - | 16 - 17 | 2096 | 95 |
AK000069.1 | - | 9 - 10 | 1242 | 95 | |
AK095861.1 | - | 17 - 18 | 2621 | 95 | |
AK296715.1 | - | 16 - 17 | 1827 | 95 | |
AK307398.1 | - | 16 - 17 | 2110 | 95 | |
AK315010.1 | - | 17 - 18 | 2046 | 95 | |
AL834482.1 | - | 11 - 12 | 1509 | 95 | |
BC000131.1 | - | 12 - 13 | 1636 | 95 | |
BC036710.2 | - | 16 - 17 | 2005 | 95 |