Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017647.3 | NP_060117.3 | 2 - 3 | 714 | 85 |
GenBank mRNA | AF327355.1 | - | 1 - 2 | 189 | 85 |
AK095861.1 | - | 2 - 3 | 714 | 85 | |
AK296715.1 | - | 2 - 3 | 113 | 85 | |
AK307398.1 | - | 1 - 2 | 203 | 85 | |
AK315010.1 | - | 2 - 3 | 139 | 85 | |
BC036710.2 | - | 1 - 2 | 98 | 85 |