Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017647.3 | NP_060117.3 | 6 - 7 | 1050 | 61 |
GenBank mRNA | AF327355.1 | - | 5 - 6 | 525 | 61 |
AK095861.1 | - | 6 - 7 | 1050 | 61 | |
AK307398.1 | - | 5 - 6 | 539 | 61 | |
AK315010.1 | - | 6 - 7 | 475 | 61 | |
BC000131.1 | - | 1 - 2 | 65 | 61 | |
BC036710.2 | - | 5 - 6 | 434 | 61 |