Assay Design



Gene Transcripts

Gene Symbol
ZNF44
Entrez Gene ID
51710
Gene Name
zinc finger protein 44
Gene Aliases
GIOT-2, KOX7, ZNF, ZNF504, ZNF55, ZNF58
Chromosome Location
Chr.19: 12224686 - 12294904 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001164276.1 NP_001157748.1 1 - 2 209 98
XM_011528062.2 XP_011526364.1 1 - 2 204 98
XM_017026871.1 XP_016882360.1 1 - 2 203 98
GenBank mRNA AK297528.1 - 1 - 2 123 98
BC001868.1 - 1 - 2 209 98
BC032246.1 - 1 - 2 132 98
BC136502.1 - 1 - 2 104 98

View Full Product Details