Assay Design
Gene Transcripts
- Gene Symbol
- ZNF44
- Entrez Gene ID
-
51710
- Gene Name
- zinc finger protein 44
- Gene Aliases
-
GIOT-2, KOX7, ZNF, ZNF504, ZNF55, ZNF58
- Chromosome Location
-
Chr.19: 12224686 - 12294904 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC032246.1 |
- |
8 - 9 |
999 |
100 |
View Full Product Details