Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001144906.1 | NP_001138378.1 | 2 - 3 | 245 | 99 |
NM_001144907.1 | NP_001138379.1 | 2 - 3 | 245 | 99 | |
NR_026709.1 | 2 - 3 | 245 | 99 | ||
GenBank mRNA | AY042238.1 | - | 2 - 3 | 127 | 99 |
AY042239.1 | - | 2 - 3 | 127 | 99 | |
AY042240.1 | - | 2 - 3 | 127 | 99 |