Assay Design



Gene Transcripts

Gene Symbol
PDZD4
Entrez Gene ID
57595
Gene Name
PDZ domain containing 4
Gene Aliases
LNX5, LU1, PDZK4, PDZRN4L
Chromosome Location
Chr.X: 153802166 - 153830567 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001303516.1 NP_001290445.1 2 - 3 401 58
NM_032512.3 NP_115901.2 2 - 3 566 58
GenBank mRNA AB103478.1 - 2 - 3 547 58
AK091444.1 - 2 - 3 292 58
AK127016.1 - 2 - 3 566 58
BC020347.1 - 1 - 2 129 58
DA097500.1 - 2 - 3 387 58

View Full Product Details