Assay Design



Gene Transcripts

Gene Symbol
PDZD4
Entrez Gene ID
57595
Gene Name
PDZ domain containing 4
Gene Aliases
LNX5, LU1, PDZK4, PDZRN4L
Chromosome Location
Chr.X: 153802166 - 153830567 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001303512.1 NP_001290441.1 6 - 7 937 99
NM_001303513.1 NP_001290442.1 7 - 8 1180 99
NM_001303514.1 NP_001290443.1 4 - 5 592 99
NM_001303515.1 NP_001290444.1 6 - 7 772 99
NM_001303516.1 NP_001290445.1 6 - 7 754 99
NM_032512.3 NP_115901.2 6 - 7 919 99
GenBank mRNA AB040877.1 - 3 - 4 257 99
AB103478.1 - 6 - 7 900 99
AK091444.1 - 6 - 7 645 99
AK123229.1 - 6 - 7 674 99
AK123822.1 - 6 - 7 772 99
AK127016.1 - 6 - 7 919 99
BC020347.1 - 5 - 6 482 99
BC117277.1 - 6 - 7 729 99
BC143459.1 - 4 - 5 383 99
BC143545.1 - 6 - 7 728 99

View Full Product Details