Assay Design



Gene Transcripts

Gene Symbol
PDZD4
Entrez Gene ID
57595
Gene Name
PDZ domain containing 4
Gene Aliases
LNX5, LU1, PDZK4, PDZRN4L
Chromosome Location
Chr.X: 153802166 - 153830567 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001303513.1 NP_001290442.1 3 - 4 824 75
GenBank mRNA AK123229.1 - 2 - 3 318 75
AK309212.1 - 3 - 4 629 75

View Full Product Details