Assay Design



Gene Transcripts

Gene Symbol
UMODL1
Entrez Gene ID
89766
Gene Name
uromodulin like 1
Gene Aliases
-
Chromosome Location
Chr.21: 42062959 - 42142996 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001004416.2 NP_001004416.2 13 - 14 2367 95
NM_001199527.1 NP_001186456.1 12 - 13 2931 95
NM_001199528.2 NP_001186457.2 13 - 14 2547 95
NM_173568.3 NP_775839.3 12 - 13 2751 95
XM_011529797.1 XP_011528099.1 10 - 11 2359 95
XM_017028506.1 XP_016883995.1 12 - 13 2746 95
XM_017028507.1 XP_016883996.1 12 - 13 5675 95
XM_017028508.1 XP_016883997.1 2 - 3 530 95
GenBank mRNA AB051810.1 - 12 - 13 2931 95
AB051811.1 - 13 - 14 2547 95
AY771619.1 - 12 - 13 2751 95
AY771620.1 - 13 - 14 2367 95

View Full Product Details