Assay Design



Gene Transcripts

Gene Symbol
UMODL1
Entrez Gene ID
89766
Gene Name
uromodulin like 1
Gene Aliases
-
Chromosome Location
Chr.21: 42062959 - 42142996 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001004416.2 NP_001004416.2 15 - 16 2681 96
NM_001199527.1 NP_001186456.1 14 - 15 3245 96
NM_001199528.2 NP_001186457.2 15 - 16 2861 96
NM_173568.3 NP_775839.3 14 - 15 3065 96
XM_011529797.1 XP_011528099.1 12 - 13 2673 96
XM_017028506.1 XP_016883995.1 14 - 15 3060 96
XM_017028507.1 XP_016883996.1 14 - 15 5989 96
XM_017028508.1 XP_016883997.1 4 - 5 844 96
GenBank mRNA AB051810.1 - 14 - 15 3245 96
AB051811.1 - 15 - 16 2861 96
AY771619.1 - 14 - 15 3065 96
AY771620.1 - 15 - 16 2681 96

View Full Product Details