Assay Design



Gene Transcripts

Gene Symbol
UMODL1
Entrez Gene ID
89766
Gene Name
uromodulin like 1
Gene Aliases
-
Chromosome Location
Chr.21: 42062959 - 42142996 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001004416.2 NP_001004416.2 16 - 17 2825 107
NM_001199527.1 NP_001186456.1 15 - 16 3389 107
NM_001199528.2 NP_001186457.2 16 - 17 3005 107
NM_173568.3 NP_775839.3 15 - 16 3209 107
XM_011529797.1 XP_011528099.1 13 - 14 2817 107
XM_017028506.1 XP_016883995.1 15 - 16 3204 107
XM_017028507.1 XP_016883996.1 15 - 16 6133 107
XM_017028508.1 XP_016883997.1 5 - 6 988 107
GenBank mRNA AB051810.1 - 15 - 16 3389 107
AB051811.1 - 16 - 17 3005 107
AK124223.1 - 2 - 3 569 107
AY771619.1 - 15 - 16 3209 107
AY771620.1 - 16 - 17 2825 107

View Full Product Details