Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001004416.2 | NP_001004416.2 | 8 - 9 | 1296 | 91 |
NM_001199527.1 | NP_001186456.1 | 8 - 9 | 1476 | 91 | |
NM_001199528.2 | NP_001186457.2 | 8 - 9 | 1476 | 91 | |
NM_173568.3 | NP_775839.3 | 8 - 9 | 1296 | 91 | |
XM_011529797.1 | XP_011528099.1 | 6 - 7 | 904 | 91 | |
XM_017028506.1 | XP_016883995.1 | 8 - 9 | 1291 | 91 | |
XM_017028507.1 | XP_016883996.1 | 8 - 9 | 4220 | 91 | |
GenBank mRNA | AB051810.1 | - | 8 - 9 | 1476 | 91 |
AB051811.1 | - | 8 - 9 | 1476 | 91 | |
AB096966.1 | - | 4 - 5 | 724 | 91 | |
AB096967.1 | - | 6 - 7 | 900 | 91 | |
AB096968.1 | - | 6 - 7 | 889 | 91 | |
AB096969.1 | - | 6 - 7 | 913 | 91 | |
AB096970.1 | - | 6 - 7 | 957 | 91 | |
AB096971.1 | - | 7 - 8 | 1069 | 91 | |
AY771619.1 | - | 8 - 9 | 1296 | 91 | |
AY771620.1 | - | 8 - 9 | 1296 | 91 |