Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001278938.1 | NP_001265867.1 | 4 - 5 | 431 | 105 |
NM_004215.4 | NP_004206.1 | 4 - 5 | 718 | 105 | |
NM_198120.2 | NP_936056.1 | 4 - 5 | 551 | 105 | |
XM_017013960.1 | XP_016869449.1 | 4 - 5 | 914 | 105 | |
GenBank mRNA | AB007619.1 | - | 4 - 5 | 599 | 105 |
AF006265.1 | - | 4 - 5 | 549 | 105 | |
AK290651.1 | - | 4 - 5 | 540 | 105 | |
AK308950.1 | - | 4 - 5 | 609 | 105 | |
AL515533.3 | - | 4 - 5 | 609 | 105 | |
AY515724.1 | - | 4 - 5 | 509 | 105 | |
AY653072.1 | - | 3 - 4 | 316 | 105 | |
BC005249.1 | - | 4 - 5 | 629 | 105 | |
BC017729.1 | - | 4 - 5 | 502 | 105 | |
BC022506.1 | - | 4 - 5 | 616 | 105 | |
BG616704.1 | - | 4 - 5 | 432 | 105 | |
CD108096.1 | - | 4 - 5 | 662 | 105 | |
CR456984.1 | - | 3 - 4 | 316 | 105 |