Assay Design



Gene Transcripts

Gene Symbol
PHF14
Entrez Gene ID
9678
Gene Name
PHD finger protein 14
Gene Aliases
-
Chromosome Location
Chr.7: 10973834 - 11188565 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014660.3 NP_055475.2 5 - 6 1657 92
NR_033435.1 4 - 5 869 92
NR_033436.1 4 - 5 869 92
XM_017012811.1 XP_016868300.1 5 - 6 1695 92
XM_017012812.1 XP_016868301.1 5 - 6 1695 92
XM_017012813.1 XP_016868302.1 5 - 6 1694 92
XM_017012814.1 XP_016868303.1 5 - 6 1694 92
XM_017012815.1 XP_016868304.1 5 - 6 1694 92
XM_017012816.1 XP_016868305.1 5 - 6 1693 92
XM_017012817.1 XP_016868306.1 4 - 5 904 92
GenBank mRNA AB018326.1 - 5 - 6 1612 92
AK294419.1 - 4 - 5 869 92
AK295461.1 - 4 - 5 852 92
BC152414.1 - 5 - 6 1612 92

View Full Product Details