Assay Design



Gene Transcripts

Gene Symbol
PHF14
Entrez Gene ID
9678
Gene Name
PHD finger protein 14
Gene Aliases
-
Chromosome Location
Chr.7: 10973834 - 11188565 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014660.3 NP_055475.2 6 - 7 1762 75
NR_033435.1 5 - 6 974 75
NR_033436.1 5 - 6 974 75
XM_017012811.1 XP_016868300.1 6 - 7 1800 75
XM_017012812.1 XP_016868301.1 6 - 7 1800 75
XM_017012813.1 XP_016868302.1 6 - 7 1799 75
XM_017012814.1 XP_016868303.1 6 - 7 1799 75
XM_017012815.1 XP_016868304.1 6 - 7 1799 75
XM_017012816.1 XP_016868305.1 6 - 7 1798 75
XM_017012817.1 XP_016868306.1 5 - 6 1009 75
GenBank mRNA AB018326.1 - 6 - 7 1717 75
AK294419.1 - 5 - 6 974 75
AK295461.1 - 5 - 6 957 75
BC152414.1 - 6 - 7 1717 75

View Full Product Details