Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014660.3 | NP_055475.2 | 9 - 10 | 2330 | 195 |
NR_033435.1 | 8 - 9 | 1542 | 195 | ||
NR_033436.1 | 8 - 9 | 1542 | 195 | ||
XM_017012811.1 | XP_016868300.1 | 9 - 10 | 2368 | 195 | |
XM_017012812.1 | XP_016868301.1 | 9 - 10 | 2368 | 195 | |
XM_017012813.1 | XP_016868302.1 | 9 - 10 | 2367 | 195 | |
XM_017012814.1 | XP_016868303.1 | 9 - 10 | 2367 | 195 | |
XM_017012815.1 | XP_016868304.1 | 9 - 10 | 2367 | 195 | |
XM_017012816.1 | XP_016868305.1 | 9 - 10 | 2366 | 195 | |
XM_017012817.1 | XP_016868306.1 | 8 - 9 | 1577 | 195 | |
GenBank mRNA | AB018326.1 | - | 9 - 10 | 2285 | 195 |
AK294419.1 | - | 8 - 9 | 1542 | 195 | |
AK295461.1 | - | 8 - 9 | 1525 | 195 | |
BC152414.1 | - | 9 - 10 | 2285 | 195 |