Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001256310.1 | NP_001243239.1 | 14 - 15 | 1908 | 148 |
GenBank mRNA | AF097492.1 | - | 14 - 15 | 1542 | 148 |
AF158555.1 | - | 15 - 16 | 1888 | 148 | |
AK001220.1 | - | 11 - 12 | 980 | 148 | |
CR749593.1 | - | 7 - 8 | 2367 | 148 |