Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001082578.1 | NP_001076047.1 | 3 - 4 | 572 | 61 |
NM_014309.2 | NP_055124.1 | 2 - 3 | 615 | 61 | |
XM_005261428.2 | XP_005261485.1 | 3 - 4 | 927 | 61 | |
XM_005261430.2 | XP_005261487.1 | 3 - 4 | 927 | 61 | |
XM_005261431.2 | XP_005261488.1 | 3 - 4 | 927 | 61 | |
XM_005261433.2 | XP_005261490.1 | 3 - 4 | 927 | 61 | |
XM_005261437.1 | XP_005261494.1 | 2 - 3 | 619 | 61 | |
XM_006724185.2 | XP_006724248.1 | 3 - 4 | 927 | 61 | |
XM_006724187.2 | XP_006724250.1 | 3 - 4 | 927 | 61 | |
XM_006724188.2 | XP_006724251.1 | 3 - 4 | 927 | 61 | |
XM_006724189.3 | XP_006724252.1 | 3 - 4 | 620 | 61 | |
XM_006724191.1 | XP_006724254.1 | 2 - 3 | 619 | 61 | |
XM_006724192.2 | XP_006724255.1 | 2 - 3 | 449 | 61 | |
XM_006724193.3 | XP_006724256.1 | 3 - 4 | 927 | 61 | |
XM_017028686.1 | XP_016884175.1 | 3 - 4 | 306 | 61 | |
XM_017028687.1 | XP_016884176.1 | 3 - 4 | 487 | 61 | |
XM_017028690.1 | XP_016884179.1 | 2 - 3 | 369 | 61 | |
XM_017028693.1 | XP_016884182.1 | 2 - 3 | 619 | 61 | |
XM_017028696.1 | XP_016884185.1 | 2 - 3 | 619 | 61 | |
GenBank mRNA | AK291460.1 | - | 2 - 3 | 496 | 61 |
AL009266.1 | - | 2 - 3 | 420 | 61 | |
DQ778625.1 | - | 3 - 4 | 460 | 61 |