Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002556.2 | NP_002547.1 | 13 - 14 | 2764 | 67 |
GenBank mRNA | AF185696.1 | - | 13 - 14 | 2764 | 67 |
AK299071.1 | - | 9 - 10 | 1208 | 67 | |
BC011581.1 | - | 13 - 14 | 2352 | 67 | |
BC063121.1 | - | 7 - 8 | 1123 | 67 | |
M86917.1 | - | 13 - 14 | 2764 | 67 |