Assay Design



Gene Transcripts

Gene Symbol
OSBP
Entrez Gene ID
5007
Gene Name
oxysterol binding protein
Gene Aliases
OSBP1
Chromosome Location
Chr.11: 59574398 - 59616144 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002556.2 NP_002547.1 1 - 1 761 61
GenBank mRNA AF185696.1 - 1 - 1 761 61
BC011581.1 - 1 - 1 349 61
M86917.1 - 1 - 1 761 61

View Full Product Details