Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001163817.1 | NP_001157289.1 | 3 - 4 | 351 | 74 |
NM_001360.2 | NP_001351.2 | 3 - 4 | 374 | 74 | |
XM_011544777.1 | XP_011543079.1 | 3 - 4 | 324 | 74 | |
GenBank mRNA | AF034544.1 | - | 3 - 4 | 295 | 74 |
AF062481.1 | - | 1 - 2 | 64 | 74 | |
AF067127.1 | - | 2 - 3 | 203 | 74 | |
AF096305.1 | - | 1 - 2 | 106 | 74 | |
AK312775.1 | - | 3 - 4 | 328 | 74 | |
BC000054.2 | - | 3 - 4 | 276 | 74 | |
BU848891.1 | - | 3 - 4 | 258 | 74 | |
DA502590.1 | - | 3 - 4 | 374 | 74 | |
KJ534821.1 | - | 1 - 2 | 101 | 74 | |
KJ534963.1 | - | 1 - 2 | 101 | 74 | |
KJ534997.1 | - | 1 - 2 | 101 | 74 | |
KU178026.1 | - | 1 - 2 | 101 | 74 | |
KU178027.1 | - | 1 - 2 | 101 | 74 | |
KU178028.1 | - | 1 - 2 | 101 | 74 | |
KU178029.1 | - | 1 - 2 | 101 | 74 |