Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000940.2 | NP_000931.1 | 1 - 2 | 102 | 73 |
GenBank mRNA | AF320003.1 | - | 1 - 2 | 77 | 73 |
AF329433.1 | - | 1 - 2 | 77 | 73 | |
AK304282.1 | - | 1 - 2 | 95 | 73 | |
AV655538.1 | - | 1 - 2 | 102 | 73 | |
BC070374.1 | - | 1 - 2 | 88 | 73 | |
CD014095.1 | - | 1 - 2 | 90 | 73 | |
CD014097.1 | - | 1 - 2 | 90 | 73 | |
CD014100.1 | - | 1 - 2 | 90 | 73 | |
CD014102.1 | - | 1 - 2 | 90 | 73 |