Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_033127.2 | NP_149118.2 | 6 - 7 | 880 | 122 |
GenBank mRNA | AB063357.1 | - | 6 - 7 | 880 | 122 |
AK314585.1 | - | 6 - 7 | 946 | 122 | |
EF125213.1 | - | 5 - 6 | 790 | 122 |