Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_033127.2 | NP_149118.2 | 8 - 9 | 1084 | 118 |
GenBank mRNA | AB063357.1 | - | 8 - 9 | 1084 | 118 |
BC009106.1 | - | 8 - 9 | 1079 | 118 | |
EF125213.1 | - | 7 - 8 | 994 | 118 |