Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_033127.2 | NP_149118.2 | 14 - 15 | 1863 | 79 |
GenBank mRNA | AB063357.1 | - | 14 - 15 | 1863 | 79 |
AK098627.1 | - | 3 - 4 | 483 | 79 | |
AK314585.1 | - | 13 - 14 | 1867 | 79 | |
EF125213.1 | - | 13 - 14 | 1773 | 79 |