Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_174952.2 | NP_777612.1 | 7 - 8 | 1024 | 154 |
XM_011531886.2 | XP_011530188.1 | 9 - 10 | 1524 | 154 | |
XM_017008049.1 | XP_016863538.1 | 7 - 8 | 1299 | 154 | |
XM_017008050.1 | XP_016863539.1 | 7 - 8 | 1257 | 154 | |
XM_017008051.1 | XP_016863540.1 | 4 - 5 | 502 | 154 | |
GenBank mRNA | BC036870.1 | - | 7 - 8 | 1024 | 154 |
BX648305.1 | - | 1 - 2 | 76 | 154 |