Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_174952.2 | NP_777612.1 | 9 - 10 | 1293 | 141 |
XM_011531886.2 | XP_011530188.1 | 11 - 12 | 1793 | 141 | |
XM_017008049.1 | XP_016863538.1 | 9 - 10 | 1568 | 141 | |
XM_017008050.1 | XP_016863539.1 | 9 - 10 | 1526 | 141 | |
XM_017008051.1 | XP_016863540.1 | 6 - 7 | 771 | 141 | |
GenBank mRNA | BC036870.1 | - | 9 - 10 | 1293 | 141 |
BX648305.1 | - | 3 - 4 | 345 | 141 |